Learn More
StatLab PathFlow PiSmart Laser Attached Lid Front Hinge Microbiopsy Cassettes
Description
- 0.38 mm pores
- Front hinge with lid attached
- Made from acetal polymer
- Specifically validated for the Pi Smart Laser Cassette Printer
- Print face features four raised pips in the corners to protect printed surface during dewaxing, while enabling the surface to be cleared of wax
- Threaded
- 2000 per case
Specifications
Specifications
| Quantity | 2000 Cassettes/Cs. |
| Color | Blue |
| Product Type | Microbiopsy Cassettes |
Made in USA