Learn More
StatLab PathFlow Connected Lid Cassettes Slotted
Catalog No. 21402133
Change view
Click to view available options
Pore Style:
Loose
Taped
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Style
21402133 Taped
21402132 Loose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21402133 Supplier StatLab Supplier No. CAS505PCT
Only null left
Cassettes slotted, one piece design with rear hinge and front tab closure
  • The 35 degree print face is designed to be compatible for printing on the Leica IP C Inkjet Cassette Printer, Primera Signature cassette printer and the Tissue-Tek SmartWrite Cassette Printer
  • These cassettes are a suitable replacement for the Unisette (M505) and Signature cassettes(M405)
  • Loose,1500 per case
Specifications
Pore Style Taped
Color Peach
Product Type Slotted Embedding Cassette