missing translation for 'onlineSavingsMsg'
Learn More

Patent Blue V, sodium salt

CAS: 20262-76-4 | C27H31N2NaO7S2 | 582.662 g/mol

$44.81 - $94.31

Chemical Identifiers

CAS 20262-76-4
Molecular Formula C27H31N2NaO7S2
Molecular Weight (g/mol) 582.662
MDL Number MFCD09608138
InChI Key PMLFOMWMYRKZRF-UHFFFAOYSA-M
Synonym m-Hydroxytetraethyldiaminotriphenylcarbinol anhydride disulfonic acid, sodium salt, Acid Blue 3
PubChem CID 9916325
IUPAC Name sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate
SMILES CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC339330050
SDS View Documents
Thermo Scientific Chemicals
339330050
5 g Glass bottle
Each for $44.81
Only null left
 
AC339330250
SDS View Documents
Thermo Scientific Chemicals
339330250
25 g Glass bottle
Each for $94.31
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20262-76-4
582.662
PMLFOMWMYRKZRF-UHFFFAOYSA-M
9916325
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+]
C27H31N2NaO7S2
MFCD09608138
m-Hydroxytetraethyldiaminotriphenylcarbinol anhydride disulfonic acid, sodium salt, Acid Blue 3
sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate

Specifications

20262-76-4
C27H31N2NaO7S2
m-Hydroxytetraethyldiaminotriphenylcarbinol anhydride disulfonic acid, sodium salt, Acid Blue 3
PMLFOMWMYRKZRF-UHFFFAOYSA-M
CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+]
582.662
582.67
Blue/Green
Crystalline Powder
80% min.
MFCD09608138
Solubility in water: soluble.
Authentic
sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate
9916325
Glass bottle
5 g
Patent blue V, sodium salt
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only