missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ PAS Staining Kit
Description
- Kit contains ready-to-use Schiff's reagent and periodic acid solution for the staining of ≈2,000 slides
- The combination of PAS staining with Alcian blue allows the additional identification of acidic mucosubstances (glycosaminoglycans)
- nuclei: blue
- acid mucosubstances: light blue
- polysaccharides: purple
- neutral mucopolysaccharides: purple
Specifications
Specifications
| Packaging | Fiber Carton |
| Quantity | 2 × 500mL |
| Storage Requirements | 15° to 25°C |
| Product Type | Stain Kit |