missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Parts and Accessories for Transferpette™ S, Transferpette™ Electronic, and Tranferpette™ Pro Single Channel Pipettes

Catalog No. 13689210
Change view
Click to view available options
Volume (Metric):
0.5 to 5 mL
1 to 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13689210 0.5 to 5 mL
13689211 1 to 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13689210 Supplier BRANDTECH™ BRAND™ Supplier No. 704652
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Replacement tip cone filters for Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro pipettes.

  • 704652: For Transferpette™ S, Transferpette™ electronic, and Transferpette™ pro, 2 mL, 2.5 mL, and 5 mL
  • 704653: For Transferpette™ S and Transferpette™ pro, 1 to 10 mL

Specifications

Description Replacement filter for BRAND Transferpette and Transferpette S, 0.5-5mL
Autoclavable Autoclavable
Disposable Reusable
For Use With Transferpette S, Transferpette Electronic, Transferpette Pro
Material Polyethylene
Type Filter
Volume (Metric) 0.5 to 5 mL
Filter Type PE
For Use With (Equipment) Single Channel Pipette, 2, 2.5, 5 mL
Quantity 25 Pk.
Temperature (English) Operating Ambient
Temperature (Metric) Operating Ambient
Show More Show Less