Learn More
Parr™ Quick Grip Electrodes For Calorimeter
For use with Parr™ calorimeter oxygen combustion bomb
$126.71 - $191.10
Specifications
For Use With (Equipment) Parr Calorimeter Oxygen Combustion Bomb
Includes Sleeve
View More Specs
Includes: Sleeve
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
0434925
View Documents Promotion Details
Parr™
PAR4A10
Straight Electrode
Pack of 2 Each for $126.71
Only null left
PAT5A10
View Documents Promotion Details
Parr™
5A10
Loop Electrode
Each for $191.10
Only null left
Description
Description
  • Available in straight and loop model
Specifications
Specifications
Parr Calorimeter Oxygen Combustion Bomb
Sleeve