missing translation for 'onlineSavingsMsg'
Learn More

Paroxetine Hydrochloride Hemihydrate 98.0+%, TCI America™

Catalog No. P19775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P19775G 5 g
P19771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P19775G Supplier TCI America Supplier No. P19775G

Chemical Identifiers

CAS 110429-35-1
Molecular Formula C38H42F2N2O7
Molecular Weight (g/mol) 676.758
MDL Number MFCD03658863
InChI Key ZXJDTEDKPXHKJZ-HBQYTBQASA-N
Synonym (3S,4R)-(-)-3-[(1,3-Benzodioxol-5-yloxy)methyl]-4-(4-fluorophenyl)piperidine Hydrochloride
PubChem CID 56829429
IUPAC Name (3S,4R)-3-(1,3-benzodioxol-5-yloxymethyl)-4-(4-fluorophenyl)piperidine;hydrate
SMILES C1CNCC(C1C2=CC=C(C=C2)F)COC3=CC4=C(C=C3)OCO4.C1CNCC(C1C2=CC=C(C=C2)F)COC3=CC4=C(C=C3)OCO4.O

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 5 g
Formula Weight 365.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Paroxetine Hydrochloride Hemihydrate
RTECSNumber TM4569320
TSCA No