missing translation for 'onlineSavingsMsg'
Learn More

Paromomycin sulfate

Catalog No. AAJ6127403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6127403 1 g
AAJ6127406 5 g
AAJ6127414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6127403 Supplier Thermo Scientific Chemicals Supplier No. J6127403
Only null left

Inhibits initiation and elongation during protein synthesis | CAS: 1263-89-4 | C23H47N5O18S | 713.706 g/mol

Paromomycin sulfate salt has been used as a reference compound in antileishmanial activity, RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex. It has also been used in cell culture applications at 100mg/L. Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Paromomycin sulfate salt has been used as a reference compound in antileishmanial activity, RNA-binding ligand and interacts with aptamer. This interaction prevents the binding (and cutting) of dicer to RNA duplex. It has also been used in cell culture applications at 100mg/L. Paromomycin inhibits the initiation and elongation steps of protein synthesis by binding to 16S ribosomal RNA. Paramomycin binds to the A site, which causes defective polypeptide chains to be produced and leads to cell death.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Chemical Identifiers

CAS 1263-89-4
Molecular Formula C23H47N5O18S
Molecular Weight (g/mol) 713.706
MDL Number MFCD00079278
InChI Key LJRDOKAZOAKLDU-JXDXLELXSA-N
PubChem CID 91972180
IUPAC Name (3S,5S,6S)-5-amino-2-(aminomethyl)-6-[(2R,4S,5S)-5-[(2R,5R,6R)-3,5-diamino-2-[(2S,3S,5S)-3-amino-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-6-hydroxycyclohexyl]oxy-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]oxyoxane-3,4-diol;sulfuric acid
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CO)O)O)N)OC3C(C(C(O3)CO)OC4C(C(C(C(O4)CN)O)O)N)O)O)N.OS(=O)(=O)O

Specifications

Type Paromomycin Sulfate
Physical Form Powder
Color White
Quantity 1 g
Optical Rotation +51° (c=5 in Water)
Beilstein 5715182
Merck Index 14,7041
Solubility Information Soluble in water
Formula Weight 713.71
Chemical Name or Material Paromomycin sulfate
Hazard Category H315-H320-H335-H360
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 215-031-7
RTECSNumber WK2320000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only