missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Parecoxib sodium salt

Chemical Identifiers

CAS 198470-85-8
Molecular Formula C19H17N2NaO4S
Molecular Weight (g/mol) 392.40
InChI Key HQPVVKXJNZEAFW-UHFFFAOYSA-M
IUPAC Name sodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
SMILES [Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC465380010
View Documents
Thermo Scientific™
465380010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC465380050
View Documents
Thermo Scientific™
465380050
5 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

198470-85-8
392.40
sodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
C19H17N2NaO4S
HQPVVKXJNZEAFW-UHFFFAOYSA-M
[Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1

Specifications

198470-85-8
1 g
[Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1
392.40
Parecoxib sodium salt
C19H17N2NaO4S
HQPVVKXJNZEAFW-UHFFFAOYSA-M
sodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
392.4
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

Suspected of damaging fertility or the unborn child.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only