missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Parecoxib sodium salt

Catalog No. AC465380010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465380010 1 g
AC465380050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465380010 Supplier Thermo Scientific Chemicals Supplier No. 465380010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 198470-85-8 | C19H17N2NaO4S | 392.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 198470-85-8
InChI Key HQPVVKXJNZEAFW-UHFFFAOYSA-M
IUPAC Name sodium [4-(5-methyl-3-phenyl-1,2-oxazol-4-yl)benzenesulfonyl](propanoyl)azanide
SMILES [Na+].CCC(=O)[N-]S(=O)(=O)C1=CC=C(C=C1)C1=C(C)ON=C1C1=CC=CC=C1

Specifications

Melting Point 273.0°C to 275.0°C
Molecular Formula C19H17N2NaO4S
Quantity 1 g
Molecular Weight (g/mol) 392.40
Formula Weight 392.4
Chemical Name or Material Parecoxib sodium salt
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

If eye irritation persists: Get medical advice/attention.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only