Learn More
Thermo Scientific Chemicals Pararosaniline chloride, pure, high purity biological stain
CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol
Supplier: Thermo Scientific Chemicals 227880250
| Quantity | 25 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 569-61-9 | |
| 323.824 | |
| JUQPZRLQQYSMEQ-UHFFFAOYSA-N | |
| 11292 | |
| 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
| C19H18ClN3 | |
| MFCD00001657 | |
| Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500 | |
| CHEBI:87663 | |
| C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl |
Specifications
| Pararosaniline chloride | |
| 569-61-9 | |
| MFCD00001657 | |
| Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol | |
| C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl | |
| 323.824 | |
| CHEBI:87663 | |
| Pure | |
| 25 g |
| 268.0°C to 270.0°C | |
| C19H18ClN3 | |
| Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500 | |
| JUQPZRLQQYSMEQ-UHFFFAOYSA-N | |
| 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride | |
| 11292 | |
| 323.83 | |
| Glass bottle |
Safety and Handling
GHS H Statement:
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
EINECSNumber : 209-321-2
RTECSNumber : CX9850100
TSCA : TSCA