missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Pararosaniline chloride, pure, high purity biological stain

Catalog No. AC227880050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227880050 5 g Glass bottle
AC227880250 25 g Glass bottle
AC227881000 100 g Glass bottle
AC227885000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC227880050 Supplier Thermo Scientific Chemicals Supplier No. 227880050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 569-61-9 | C19H18ClN3 | 323.824 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 569-61-9
Molecular Formula C19H18ClN3
Molecular Weight (g/mol) 323.824
MDL Number MFCD00001657
InChI Key JUQPZRLQQYSMEQ-UHFFFAOYSA-N
Synonym Basic Red 9, monohydrochloride, Basic Fuchsin, C.I. 42500
PubChem CID 11292
ChEBI CHEBI:87663
IUPAC Name 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl

Specifications

Melting Point 268.0°C to 270.0°C
Solubility Information Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol
Formula Weight 323.83
Grade Pure
Packaging Glass bottle
Quantity 5 g
Chemical Name or Material Pararosaniline chloride
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause cancer.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-321-2
RTECSNumber CX9850100
TSCA TSCA

RUO – Research Use Only