Learn More
Thermo Scientific Chemicals Pararosaniline chloride, 96%, pure
CAS: 569-61-9 | C19H17N3·HCl | 323.83 g/mol
Supplier: Thermo Scientific Chemicals 416800050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ideal for analysis by ICP-MS and high-resolution ICP-MS.
- Evaluated for 65 metals at low-level ppt concentrations.
- Manufactured, purified, and packaged in a Class 1000 clean room.
- Packaged in specially cleaned Teflon FEP for PFA bottles to maintain product integrity and to eliminate metal leaching from glass bottles.
- Three year shelf life from time of manufacture - available on COA
Specifications
| Pararosaniline chloride | |
| 569-61-9 | |
| Crystalline Powder | |
| 97.5% min. (HPLC) | |
| basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin | |
| JUQPZRLQQYSMEQ-UHFFFAOYSA-N | |
| 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride | |
| 11292 | |
| 323.83 | |
| Pure | |
| Glass bottle |
| 268.0°C to 270.0°C | |
| Green | |
| 5 g | |
| C19H17N3·HCl | |
| Solubility in water: 10g/L (25°C). Other solubilities: 65g/L ethanol | |
| C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl | |
| 323.83 | |
| CHEBI:87663 | |
| 95% | |
| Authentic |
Chemical Identifiers
| 569-61-9 | |
| 323.83 | |
| basic red 9, pararosaniline hydrochloride, parafuchsin, pararosaniline chloride, parafuchsine, p-fuchsin, basic fuchsin, ci basic red 9, c.i. basic red 9, parafuksin | |
| CHEBI:87663 | |
| C1=CC(=N)C=CC1=C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N.Cl |
| C19H17N3·HCl | |
| JUQPZRLQQYSMEQ-UHFFFAOYSA-N | |
| 11292 | |
| 4-[(4-aminophenyl)-(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride |
Safety and Handling
GHS H Statement:
May cause cancer.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 209-321-2
RUO – Research Use Only