missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde 90.0+%, TCI America™

Supplier:  TCI America P001825G

 View more versions of this product

Catalog No. P001825G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Crystalline Powder
30525-89-4
25 g
MFCD00133991
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
C=O
30.026
CHEBI:16842
Paraformaldehyde
White
CH2O
2213
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
≥90.0% (T)

Chemical Identifiers

30525-89-4
30.026
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00133991
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1941

RTECSNumber : RV0540000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions