missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde 90.0+%, TCI America™

Catalog No. P0018500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0018500G 500 g
P001825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0018500G Supplier TCI America Supplier No. P0018500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
MDL Number MFCD00133991
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Physical Form Crystalline Powder
Color White
Quantity 500 g
UN Number 2213
Percent Purity ≥90.0% (T)
Chemical Name or Material Paraformaldehyde
EINECSNumber (9)-1941
RTECSNumber RV0540000
TSCA Yes