missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paraformaldehyde, Purified, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Pail |
| Quantity | 2.5 kg |
| Percent Purity | 90% min. |
| Grade | Purified |
| Physical Form | Powder or Waxy Solid |
| Chemical Name or Material | Paraformaldehyde |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.