missing translation for 'onlineSavingsMsg'
Learn More

Paraformaldehyde, 90%, pure

Catalog No. AC169650100
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic Bucket
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169650100 10 kg Plastic Bucket
AC169650010 1 kg Plastic bottle
AC169650025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC169650100 Supplier Thermo Scientific Chemicals Supplier No. 169650100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 30525-89-4 | CH2O | 30.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30525-89-4
Molecular Formula CH2O
Molecular Weight (g/mol) 30.026
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Melting Point 120.0°C to 170.0°C
Color White to off-white
pH 3.5 to 5 (10% suspension)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 89.5 to 93%
Packaging Plastic Bucket
Linear Formula (CH2O)n
Quantity 10 kg
Beilstein 01,566
Merck Index 15,7129
Solubility Information Solubility in water: sparingly soluble. Other solubilities: the higher polymers are insoluble in water, insoluble in alcohol and ether
Percent Purity 90%
Grade Pure
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Paraformaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Harmful if inhaled.<br
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF

RUO – Research Use Only