missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Paraformaldehyde
Description
Paraformaldehyde
Specifications
Specifications
| CAS | 30525-89-4 |
| Melting Point | 120°C to 170°C |
| Density | 0.88 g/mL at 25°C |
| Molecular Formula | CH2O ]n |
| Linear Formula | HO(CH2O)nH |
| MDL Number | MFCD00133991 |
| Quantity | 500 g |
| Synonym | Polyoxymethylene |
| Molecular Weight (g/mol) | 30.03 g/mol |