missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Paraformaldehyde

Catalog No. 4412441KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4412441KG 1 kg
5000031040 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 4412441KG Supplier Sigma Aldrich Supplier No. 4412441KG*
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Paraformaldehyde

Specifications

CAS 30525-89-4
Melting Point 120°C to 170°C (lit.)
Density 0.88 g/mL (at 25°C (literature))
Molecular Formula CH2O]n
Linear Formula HO(CH2O)nH
MDL Number MFCD00133991
Quantity 1 kg
Synonym Polyoxymethylene
Molecular Weight (g/mol) 30.03 (as monomer)
Percent Purity 95%