missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paraformaldehyde, 16% w/v aq. soln., methanol free

Catalog No. p-7058859
Change view
Click to view available options
Quantity:
2 x 10 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA433689L 2 x 10 mL
AA433689M 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA433689L Supplier Thermo Scientific Chemicals Supplier No. 0433689L
Only null left

Paraformaldehyde, CH2O, CAS Number-30525-89-4, methylene oxide, oxomethane, paraform, oxymethylene, formalin, formol, methyl aldehyde, paraformaldehyde, formic aldehyde, methanal, 10 x 10mL, CHEBI:16842, Hazard Class: 9; Packaging Group: III, 200-001-8, H312+H332-H315-H317-H319-H335-H341-H350

Paraformaldehyde is used as a disinfectant, hardening agent and waterproofing agent. It is also used to prepare adhesives, resins and dentistry as an antiseptic and contraceptive. It is also employed as a thermoplastic. In addition, it is used in the preparation of formalin fixatives for tissues or cells during the samples subjected to florescence studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon

Chemical Identifiers

InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging Ampule
Quantity 2 x 10 mL
MDL Number MFCD00133991
Merck Index 14,7025
Solubility Information Soluble in chlorophenol and many organic solvents. Insoluble in ether and alcohol.
Physical Form Liquid
Chemical Name or Material Paraformaldehyde, Methanol free
Hazard Category H312+H332-H315-H317-H319-H335-H341-H350
Hazard Statement GHS H Statement
H351-H315-H319-H317
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P301+P330+P331-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 200-001-8
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only