Learn More
Mopec™ Paraffin Wax Dispenser
Description
- Designed to take up minimal bench-top space
- Digital temperature control
- 15 L dispenser has 2 separate tanks with independent heating controls
- Rapid heating system ensures system is ready to use in less time
- Heated tap prevents wax buildup
- Non-drip lever with three positions
- Double-layered tanks provide internal temperature control and ensures outer surfaces are safe to touch
- Anti-microbial coating to help prevent bacteria and virus growth
Specifications
Specifications
| Capacity (Metric) | 15 L |
| Description | Paraffin Wax Dispenser |