Learn More
Mopec™ Paraffin Wax Dispenser
Catalog No. 22445493
Change view
Click to view available options
Capacity (Metric):
7.5 L
15 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22445493 7.5 L
22445660 15 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22445493 Supplier Mopec™ Supplier No. BK575
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Paraffin Wax Dispenser is designed to take up minimal bench-top space while providing a rapid heating system that produces melted paraffin wax in only 30 minutes.
  • Designed to take up minimal bench-top space
  • Digital temperature control
  • 15 L dispenser has 2 separate tanks with independent heating controls
  • Rapid heating system ensures system is ready to use in less time
  • Heated tap prevents wax buildup
  • Non-drip lever with three positions
  • Double-layered tanks provide internal temperature control and ensures outer surfaces are safe to touch
  • Anti-microbial coating to help prevent bacteria and virus growth
Specifications
Capacity (Metric) 7.5 L
Description Paraffin Wax Dispenser