missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Paraffin wax

Catalog No. 4116631KG
Encompass
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4116631KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4116631KG Supplier Sigma Aldrich Supplier No. 4116631KG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Paraffin wax

Specifications

CAS 8002-74-2
Melting Point ≥65°C (ASTM D 87)
MDL Number MFCD00132833
Quantity 1 kg