missing translation for 'onlineSavingsMsg'
Learn More

Paraffin Oil Light, MilliporeSigma™

Catalog No. MPX00471
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MPX00471 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPX00471 Supplier Technidata Supplier No. US1PX0047/1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 8012-95-1
Molecular Formula MFCD00131611
Molecular Weight (g/mol) 0.00
MDL Number MFCD00131611
InChI Key FFNDMZIBVDSQFI-UHFFFAOYSA-N
Synonym delphinidin chloride, delphinidin, delphinidine, delphinidol, ephdine, delfinidol chloride, unii-em6md4aehe, ccris 2518, em6md4aehe, 3,5,7-trihydroxy-2-3,4,5-trihydroxyphenyl benzopyrylium chloride
PubChem CID 68245
ChEBI CHEBI:38701
IUPAC Name 2-(3,4,5-trihydroxyphenyl)chromenylium-3,5,7-triol;chloride
SMILES *

Specifications

Density 0.85g/cm3 (20°C)
Ignition Point >300°C
Packaging Glass Bottle
Linear Formula Indeterminate
Quantity 500 mL
Vapor Pressure 0.0001 hPa (20°C)
Chemical Name or Material Paraffin Oil, For Atomic Absorption Spectroscopy
Recommended Storage Store at +15°C to +25°C