missing translation for 'onlineSavingsMsg'
Learn More

Paraffin, NF, Spectrum™ Chemical

Catalog No. p-7157933
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604991 2.5 kg Poly Pail
18604989 6 x 500 g Amber Glass Bottle
18604990 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18604991 Supplier Spectrum Chemical Mfg Cor Supplier No. P17622.5KG
Only null left

P1762, 8002-74-2

Spectrum™ Chemical Paraffin, NF is used as an ointment base in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 8002-74-2
Molecular Formula C21H27NO3
Molecular Weight (g/mol) 341.45
InChI Key JWHAUXFOSRPERK-UHFFFAOYNA-N
IUPAC Name 1-{2-[2-hydroxy-3-(propylamino)propoxy]phenyl}-3-phenylpropan-1-one
SMILES CCCNCC(O)COC1=CC=CC=C1C(=O)CCC1=CC=CC=C1

Specifications

Alkalinity To pass test
Acidity To pass test
CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Grade NF