Learn More
Simport™ Scientific Paraffin Block Mailer
Catalog No. 22045043
Change view
Click to view available options
Quantity:
50 Pk.
Catalog No. Quantity
22045043 50 Pk.
1 options
Catalog No. 22045043 Supplier Simport™ Scientific Supplier No. M4776
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Paraffin Block Mailer is a transport container designed especially for paraffin blocks.
  • Made of PVC
  • Transparent for easy viewing of contents
  • Used for handling and shipping up to 6 paraffin blocks
  • Suitable for all regular models of tissue and biopsy cassettes
  • Easy to close cover attached
Specifications
Quantity 50 Pk.
Type Block Mailer
Dimensions (LxWxH) 134 x 141 x 29 mm
Capacity 6 Blocks
Material PVC