Learn More
Epredia™ Para-Tray
Collect paraffin shavings in front of a microtome with the Epredia™ Para-Tray.
Supplier: Epredia™ 58
Description
- Stainless-steel tray
- Dimensions: 11 x 6 x 1.25in. (28.6 x 15.2 x 3.2cm)
Specifications
| Para-Tray | |
| Stainless Steel |