missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR 98.0+%, TCI America™
$72.70 - $279.60
Chemical Identifiers
| CAS | 1141-59-9 |
|---|---|
| Molecular Formula | C11H9N3O2 |
| Molecular Weight (g/mol) | 215.212 |
| MDL Number | MFCD00006256 |
| InChI Key | VLCAILLZPUINNF-LCYFTJDESA-N |
| Synonym | 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl |
| PubChem CID | 5474737 |
| IUPAC Name | (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one |
| SMILES | C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O |
Chemical Identifiers
| 1141-59-9 | |
| 215.212 | |
| VLCAILLZPUINNF-LCYFTJDESA-N | |
| 5474737 | |
| C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O |
| C11H9N3O2 | |
| MFCD00006256 | |
| 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl | |
| (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one |
Specifications
| 1141-59-9 | |
| MFCD00006256 | |
| 2811 | |
| VLCAILLZPUINNF-LCYFTJDESA-N | |
| (4Z)-3-hydroxy-4-(pyridin-2-ylhydrazinylidene)cyclohexa-2,5-dien-1-one | |
| 5474737 | |
| ≥98.0% (HPLC,T) | |
| PAR [=4-(2-Pyridylazo)resorcinol] [Spectrophotometric reagent for transition metals] |
| C11H9N3O2 | |
| 1 g | |
| 4-2-pyridylazo resorcinol, 4-2-pyridlyazo resorcinol, 4-2-pyridylazo-2-resorcinol, resorcinol, 4-2-pyridylazo, 1-2-pyridylazo resorcinol, unii-n68t40h95t, 1,3-benzenediol, 4-2-pyridinylazo, 4-pyridin-2-yldiazenyl benzene-1,3-diol, 4-2-pyridylazo-1,3-benzenediol, 1,3-benzenediol, 4-2-2-pyridinyl diazenyl | |
| C1=CC=NC(=C1)NN=C2C=CC(=O)C=C2O | |
| 215.212 | |
| 215.21 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes