Learn More
Bullard™ PAPR System Breathing Tubes for Loose-Fitting Facepieces
Catalog No. 296358319
Change view
Click to view available options
Size:
Regular
X-Long
X-Small
Length (English):
26 in.
38 in.
Length (Metric):
66.04 cm
83.82 cm
96.5 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Length (Metric)
296358319 X-Small 26 in. 66.04 cm
296358318 Regular - 83.82 cm
296359914 X-Long 38 in. 96.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 296358319 Supplier Bullard™ Supplier No. PA20LFBTXS
Only null left
Breathing tube for use with PA20 and PA30 series powered air-purifying respirator (PAPR) systems.
  • Breathing tube components/replacement parts to build PAPR replace parts for an existing system
  • For use with PA20 and PA30 loose-fitting facepieces
  • Choose from three lengths:
    • Extra small
    • Standard
    • Extra long
Specifications
Product Type Breathing Tube
Size X-Small
For Use With (Equipment) PA20 or PA30 Loose-Fitting Facepieces
Includes Hose clamp
Model PA20LFBTXS
Quantity 1 Ea.
Length (English) 26 in.
Length (Metric) 66.04 cm