Electron Microscopy Sciences PAPER POINTS SELECTION 2
Catalog No. 50930669
Machine rolled paper with a tapered end which offers good water absorbency. Assists you in the removal of surplus water from the knife boat, on the block face, and from the grids.
Catalog No.
50930669
Supplier
Electron Microscopy SciencesSupplier No.
7101102
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Machine rolled paper with a tapered end which offers good water absorbency. Assists you in the removal of surplus water from the knife boat, on the block face, and from the grids.