missing translation for 'onlineSavingsMsg'
Learn More

Sigma Diagnostics™ Papanicolaou Stains

Catalog No. 01288057
Change view
Click to view available options
Quantity:
1 L
Type:
OG-6 Papanicolaou Stain
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
01288057 1 L OG-6 Papanicolaou Stain
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01288057 Supplier Sigma Diagnostics™ Supplier No. HT401321L
Only null left

For routine diagnostic cytology to aid in the identification and classification of exfoliative cells

  • Orange G (certified), 0.3% and phosphotungstic acid, 0.015%, in denatured alcohol

Specifications

Color Orange
Type OG-6 Papanicolaou Stain
Format Solution
For Use With (Application) Characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic component
Quantity 1 L
Storage Requirements 18-26°C