missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Papanicolaou Stain, Gill's Modified OG-6 Formula, Ricca Chemical
Counterstain for use with Hematoxylin Stains in Papanicolaou Staining
$252.20 - $252.20
Specifications
| Chemical Name or Material | Papanicolaou Stain |
|---|---|
| Name Note | Gill's Modified OG-6 Formula |
| CAS | 64-17-5,7732-18-5,67-56-1,1936-15-8,12501-23-4 |
| CAS Min % | 84.0,14.23,0.24,0.02 |
| CAS Max % | 87.0,14.82,0.25,0.02 |
Specifications
| Papanicolaou Stain | |
| 64-17-5,7732-18-5,67-56-1,1936-15-8,12501-23-4 | |
| 87.0,14.82,0.25,0.02 | |
| Natural Poly Bottle | |
| 5 |
| Gill's Modified OG-6 Formula | |
| 84.0,14.23,0.24,0.02 | |
| Laboratory | |
| Orange | |
| Liquid |