Learn More
Epredia™ Papanicolaou EA Stains
Control the color hues and intensities of your work with Epredia™ Papanicolaou Stain EAs, useful for both gynecological and non-gynecological staining.
$333.03 - $649.29
| Catalog Number | Mfr. No. | Quantity | Type | Price | Qty | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Type | Price | Qty | ||||
|
22-050-314
|
Epredia™
75411 |
1 gal. | EA-36 Papanicolaou Stain |
Each for $649.29
|
|
||||
|
22-050-210
|
Epredia™
75511 |
1 gal. | EA-50 Papanicolaou Stain |
Each for $333.03
|
|
||||
|
22-050-212
|
Epredia™
75611 |
1 gal. | EA-65 Papanicolaou Stain |
Each for $371.06
|
|
||||
Description
- EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
- EA-65 is used with OG-6 for non-gynecological staining.
- Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.