missing translation for 'onlineSavingsMsg'
Learn More

Panthenol, USP, 99-102%, Spectrum™ Chemical

Catalog No. 18604967
Change view
Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604967 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604967 Supplier Spectrum Chemical Mfg Cor Supplier No. P191025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1910, 16485-10-2, C9H19NO4

Spectrum™ Chemical Panthenol, USP is used as a topical moisturizer and lubricant to treat skin diorders. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 16485-10-2
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00002944
InChI Key SNPLKNRPJHDVJA-UHFFFAOYNA-N
IUPAC Name 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)C(O)C(=O)NCCCO

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 64.5-68.5°C
Packaging Amber Glass Bottle
Quantity 25 g
Specific Rotation -0.05° to +0.05°
Percent Purity 99 to 102%
Grade USP