missing translation for 'onlineSavingsMsg'
Learn More

Panthenol, USP, 99-102%, Spectrum™ Chemical

Catalog No. 18604972
Change view
Click to view available options
Quantity:
100 g
500 g
12 kg
20 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604972 20 kg Fiber Drum
18604968 100 g Amber Glass Bottle
18604969 500 g Amber Glass Bottle
18604971 12 kg Poly Pail
18604970 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604972 Supplier Spectrum Chemical Mfg Cor Supplier No. P191020KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P1910, 16485-10-2, C9H19NO4

Spectrum™ Chemical Panthenol, USP is used as a topical moisturizer and lubricant to treat skin diorders. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 16485-10-2
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD00002944
InChI Key SNPLKNRPJHDVJA-UHFFFAOYNA-N
IUPAC Name 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
SMILES CC(C)(CO)C(O)C(=O)NCCCO

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Melting Range 64.5-68.5°C
Packaging Fiber Drum
Quantity 20 kg
Specific Rotation -0.05° to +0.05°
Percent Purity 99 to 102%
Grade USP