missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Panthenol, USP, 99-102%, Spectrum™ Chemical
P1910, 16485-10-2, C9H19NO4
Supplier: Spectrum Chemical Mfg Cor P191025KGBL
Description
Spectrum™ Chemical Panthenol, USP is used as a topical moisturizer and lubricant to treat skin diorders. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 16485-10-2 | |
| 0.001 | |
| 64.5-68.5°C | |
| C9H19NO4 | |
| 25 kg | |
| SNPLKNRPJHDVJA-UHFFFAOYNA-N | |
| 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide | |
| 99 to 102% |
| 1 | |
| 0.005 | |
| Poly Pail | |
| MFCD00002944 | |
| -0.05° to +0.05° | |
| CC(C)(CO)C(O)C(=O)NCCCO | |
| 205.25 | |
| USP |
Chemical Identifiers
| 16485-10-2 | |
| 205.25 | |
| SNPLKNRPJHDVJA-UHFFFAOYNA-N | |
| CC(C)(CO)C(O)C(=O)NCCCO |
| C9H19NO4 | |
| MFCD00002944 | |
| 2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide |