Learn More
Selleck Chemical Llc Panobinostat (LBH589) 200mg 404950-80-7 NVP-LBH589
Supplier: Selleck Chemicals Co Ltd S1030200MG
Panobinostat (LBH589, NVP-LBH589) is a novel broad-spectrum HDAC inhibitor with IC50 of 5 nM in a cell-free assay. Panobinostat (LBH589) induces autophagy and apoptosis. Panobinostat effectively disrupts HIV latency in vivo. Phase 3. *For Research & Development use only. Product is not intended for drug, household, or human consumption.