missing translation for 'onlineSavingsMsg'
Learn More

Pancreatin U.S.P., MP Biomedicals™

Catalog No. p-4404470
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10255780 100 g
ICN10255791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Catalog No. ICN10255780 missing translation for 'mfr' MP Biomedicals Inc missing translation for 'supplierNo' 0210255780
Only null left

Chemical Identifiers

CAS 8049-47-6
Synonym Diastase vera

Specifications

Color Tan to White
Quantity 100 g
Physical Form Powder
Chemical Name or Material Pancreatin