missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Pancreatic Polypeptide/PP Peptide

Catalog No. NB804825
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB804825 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB804825 Supplier R&D Systems™ Supplier No. NB1001793PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems.

Specifications

Gene ID (Entrez) 5539
Species Human
Product Line Novus
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
For Use With (Application) This Peptide is Useful as a Blocking Peptide for NB100-1793
Gene Alias Obinepitide, pancreatic polypeptide (ppy), pancreatic polypeptide PNPPP, pancreatic polypeptide Y, pancreatic prohormone
Gene Symbol PPY
Quantity 0.1 mg
Research Category Stem Cell Markers