missing translation for 'onlineSavingsMsg'
Learn More

PAN 98.0+%, TCI America™

Catalog No. A50001G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50001G 1 g
A50005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50001G Supplier TCI America Supplier No. A50001G

Chemical Identifiers

CAS 85-85-8
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.273
MDL Number MFCD00004071
InChI Key RAXUMGMWXZYADR-OBGWFSINSA-N
Synonym 1-2-pyridylazo-2-naphthol, unii-095b53y3xv, pan indicator, 2-naphthalenol, 1-2-pyridinylazo, 1e-1-pyridin-2-ylhydrazinylidene naphthalen-2-one, 1-2-pyridylazo-2-hydroxynaphthalene, pan van, 1-2-pyridyldiazenyl naphthalen-2-ol, 1-pyridin-2-azo-2-naphthol, 2-naphthalenol, 1-2-2-pyridinyl diazenyl
PubChem CID 5376264
IUPAC Name (1E)-1-(pyridin-2-ylhydrazinylidene)naphthalen-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=CC=N3

Specifications

Melting Point 142°C
Quantity 1 g
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material PAN [=1-(2-Pyridylazo)-2-naphthol] [Metal indicator and spectrophotometric reagent for transi