missing translation for 'onlineSavingsMsg'
Learn More

PAN, 0.1% (w/v) Indicator Solution in Methanol, Spectrum™ Chemical

Catalog No. 18604958
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604958 500 mL Amber Glass Bottle
18604957 100 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18604958 Supplier Spectrum Chemical Mfg Cor Supplier No. P358500ML
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

P-358, 85-85-8

Spectrum™ Chemical PAN, 0.1 Percent (w/v) Indicator Solution in Methanol

Chemical Identifiers

CAS 85-85-8 , 67-56-1
Molecular Formula C15H11N3O
Molecular Weight (g/mol) 249.27
InChI Key RAXUMGMWXZYADR-SDXDJHTJSA-N
IUPAC Name (1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
SMILES O=C1C=CC2=CC=CC=C2\C1=N\NC1=CC=CC=N1

Specifications

CAS Max % 1, 0.999
Boiling Point 64°C / 147.2°F
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Chemical Solution
Physical Form Liquid
Chemical Name or Material PAN, 0.1 Percent (w/v) Indicator Solution in Methanol