missing translation for 'onlineSavingsMsg'
Learn More

Pamoic Acid Disodium Salt Monohydrate 98.0+%, TCI America™

Catalog No. p-7134921
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P105925G 25 g
P1059250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P105925G Supplier TCI America Supplier No. P105925G
Only null left

Chemical Identifiers

CAS 6640-22-8
Molecular Formula C23H14Na2O6
Molecular Weight (g/mol) 432.339
MDL Number MFCD00036183
InChI Key YGLLICRFEVEWOZ-UHFFFAOYSA-L
Synonym sodium pamoate, pamoic acid disodium salt, disodium pamoate, unii-rpo7lte47e, disodium 4,4'-methylenebis 3-hydroxy-2-naphthoate, rpo7lte47e, disodium methylenebis 2-hydroxy-3-naphthoate, 2-naphthalenecarboxylic acid, 4,4'-methylenebis 3-hydroxy-, disodium salt, sodium 4,4'-methylenebis 3-hydroxy-2-naphthoate, embonic acid disodium salt
PubChem CID 54676156
IUPAC Name disodium;3-carboxy-1-[(3-carboxy-2-oxidonaphthalen-1-yl)methyl]naphthalen-2-olate
SMILES C1=CC=C2C(=C1)C=C(C(=C2CC3=C(C(=CC4=CC=CC=C43)C(=O)O)[O-])[O-])C(=O)O.[Na+].[Na+]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 432.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Pamoic Acid Disodium Salt Monohydrate