missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palonosetron Hydrochloride 98.0+%, TCI America™
Supplier: TCI America P20511G
Specifications
| Palonosetron Hydrochloride | |
| 307°C | |
| MFCD00939524 | |
| OLDRWYVIKMSFFB-KPVRICSOSA-N | |
| hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0âµ,¹³]trideca-1(13),9,11-trien-2-one chloride | |
| 6918303 | |
| 332.87 | |
| Crystalline Powder |
| 135729-62-3 | |
| C19H25ClN2O | |
| 1 g | |
| [H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2 | |
| 332.87 | |
| CHEBI:85157 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 135729-62-3 | |
| 332.87 | |
| OLDRWYVIKMSFFB-KPVRICSOSA-N | |
| CHEBI:85157 | |
| [H+].[Cl-].O=C1N(C[C@@H]2CCCC3=CC=CC1=C23)[C@@H]1CN2CCC1CC2 |
| C19H25ClN2O | |
| MFCD00939524 | |
| 6918303 | |
| hydrogen (5R)-3-[(3S)-1-azabicyclo[2.2.2]octan-3-yl]-3-azatricyclo[7.3.1.0âµ,¹³]trideca-1(13),9,11-trien-2-one chloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator