missing translation for 'onlineSavingsMsg'
Learn More

Palmityl palmitate, 96%

CAS: 540-10-3 | C32H64O2 | 480.86 g/mol

Supplier:  Thermo Scientific Chemicals H5928306

 View more versions of this product

Catalog No. AAH5928306


Only null left

Description

Description

It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.

Solubility
Soluble in hot acetone.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
Specifications

Specifications

Palmityl palmitate
55°C to 56°C
CH3(CH2)14CO2(CH2)15CH
5 g
Soluble in hot acetone.
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
480.86
CHEBI:75584
96%
540-10-3
C32H64O2
MFCD00053739
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PXDJXZJSCPSGGI-UHFFFAOYSA-N
hexadecyl hexadecanoate
10889
480.85

Chemical Identifiers

540-10-3
480.86
PXDJXZJSCPSGGI-UHFFFAOYSA-N
10889
hexadecyl hexadecanoate
C32H64O2
MFCD00053739
cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
CHEBI:75584
CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 208-736-6

RTECSNumber : RT4750000

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only