missing translation for 'onlineSavingsMsg'
Learn More

Palmityl palmitate, 96%

Catalog No. AAH5928303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5928303 1 g
AAH5928306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5928303 Supplier Thermo Scientific Chemicals Supplier No. H5928303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 540-10-3 | C32H64O2 | 480.86 g/mol

It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as emollients, masking agents and skin conditioning and a glosser and thickener for creams. It improves emulsion texture and stability and gives structure to cosmetic sticks. Used in skin and hair applications.

Solubility
Soluble in hot acetone.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.

Chemical Identifiers

CAS 540-10-3
Molecular Formula C32H64O2
Molecular Weight (g/mol) 480.86
MDL Number MFCD00053739
InChI Key PXDJXZJSCPSGGI-UHFFFAOYSA-N
Synonym cetyl palmitate, hexadecyl palmitate, palmityl palmitate, cetin, hexadecanoic acid, hexadecyl ester, standamul 1616, hexadecanoic acid hexadecyl ester, hexadecanyl hexadecanoate, n-hexadecyl hexadecanoate, palmitic acid palmityl ester
PubChem CID 10889
ChEBI CHEBI:75584
IUPAC Name hexadecyl hexadecanoate
SMILES CCCCCCCCCCCCCCCCOC(=O)CCCCCCCCCCCCCCC

Specifications

Melting Point 55°C to 56°C
Linear Formula CH3(CH2)14CO2(CH2)15CH
Quantity 1 g
Solubility Information Soluble in hot acetone.
Formula Weight 480.85
Percent Purity 96%
Chemical Name or Material Palmityl palmitate
EINECSNumber 208-736-6
RTECSNumber RT4750000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only