missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Palmitoylethanolamide, 98%
$560.16
Chemical Identifiers
| CAS | 544-31-0 |
|---|---|
| Molecular Formula | C18H37NO2 |
| Synonym | PEA; N-(2-Hydroxyethyl)hexadecanamide |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ67473MF
|
Thermo Scientific Chemicals
J67473MF |
50 mg |
Each for $560.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 544-31-0 | |
| C18H37NO2 | |
| PEA; N-(2-Hydroxyethyl)hexadecanamide |
Specifications
| White | |
| 544-31-0 | |
| 299.50 | |
| 97°C to 98°C | |
| Solid | |
| CCCCCCCCCCCCCCCC(=O)NCCO | |
| 299.50 |
| 2°C to 8°C | |
| C18H37NO2 | |
| PEA; N-(2-Hydroxyethyl)hexadecanamide | |
| Palmitoylethanolamide, 98% | |
| HXYVTAGFYLMHSO-UHFFFAOYSA-N | |
| N-(2-hydroxyethyl)hexadecanamide | |
| 98% |
Safety and Handling
EINECSNumber : 208-867-9
Recommended Storage : Keep cold
RUO – Research Use Only