missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palmitic Anhydride 96.0+%, TCI America™
Supplier: TCI America P000825G
Specifications
| Palmitic Anhydride | |
| 64°C | |
| C32H62O3 | |
| 25 g | |
| palmitic anhydride, hexadecanoic anhydride, palmitic acid anhydride, palmiticanhydride, hexadecanoic acid, anhydride, unii-33jhb8gs96, hexadecanoic acid, 1,1'-anhydride, bispalmitic anhydride, n-hexadecanoic anhydride, hexadecanoic anhydride # | |
| CCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCC | |
| 494.845 | |
| 494.85 | |
| Crystalline Powder |
| 623-65-4 | |
| White | |
| MFCD00008992 | |
| 3261 | |
| QWZBEFCPZJWDKC-UHFFFAOYSA-N | |
| hexadecanoyl hexadecanoate | |
| 69339 | |
| ≥96.0% (GC,T) |
Chemical Identifiers
| 623-65-4 | |
| 494.845 | |
| QWZBEFCPZJWDKC-UHFFFAOYSA-N | |
| 69339 | |
| CCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCC |
| C32H62O3 | |
| MFCD00008992 | |
| palmitic anhydride, hexadecanoic anhydride, palmitic acid anhydride, palmiticanhydride, hexadecanoic acid, anhydride, unii-33jhb8gs96, hexadecanoic acid, 1,1'-anhydride, bispalmitic anhydride, n-hexadecanoic anhydride, hexadecanoic anhydride # | |
| hexadecanoyl hexadecanoate |
Safety and Handling
TSCA : Yes