missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palladium wire, 0.25mm (0.01in) dia, 99.97% (metals basis)
CAS: 7440-05-3 | Pd | 106.42 g/mol
$110.33 - $110.33
Chemical Identifiers
| CAS | 5-3-7440 |
|---|---|
| Molecular Formula | Pd |
| Molecular Weight (g/mol) | 106.42 |
| MDL Number | MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 |
| InChI Key | KDLHZDBZIXYQEI-UHFFFAOYSA-N |
| Synonym | black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon |
| PubChem CID | 23938 |
| ChEBI | CHEBI:33363 |
| IUPAC Name | palladium |
| SMILES | [Pd] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 5-3-7440 | |
| 106.42 | |
| KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
| 23938 | |
| palladium |
| Pd | |
| MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
| black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
| CHEBI:33363 | |
| [Pd] |
Specifications
| 5-3-7440 | |
| Odorless | |
| Wire | |
| MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 | |
| KDLHZDBZIXYQEI-UHFFFAOYSA-N | |
| palladium | |
| 23938 | |
| 99.97% | |
| (metals basis) |
| 30 cm | |
| ≈0.6g/m | |
| Pd | |
| black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon | |
| [Pd] | |
| 106.42 | |
| CHEBI:33363 | |
| Palladium wire, 0.25mm (0.01 in.) dia. |
Safety and Handling
EINECSNumber : 231-115-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only