missing translation for 'onlineSavingsMsg'
Learn More

Palladium wire, 0.127mm (0.005in) dia, 99.9% (metals basis)

Catalog No. AA10277G2
Change view
Click to view available options
Quantity:
2 m
10 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10277G2 2 m
AA10277G6 10 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10277G2 Supplier Thermo Scientific Chemicals Supplier No. 010277G2

CAS: 7440-05-3 | Pd | 106.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Quantity 2 m
Odor Odorless
Weight ≈0.15g/m
Form Wire
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Palladium wire, 0.127mm (0.005 in.) dia.
EINECSNumber 231-115-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only