missing translation for 'onlineSavingsMsg'
Learn More

Palladium Silver foil, 0.025mm (0.001in) thick

Catalog No. AA45003FI
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45003FI 50 x 50 mm
AA45003FF 25 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45003FI Supplier Thermo Scientific Chemicals Supplier No. 045003FI

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Palladium Silver foil
Name Note 0.025mm (0.001 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pd:Ag; 77:23 wt%
Quantity 50 x 50 mm
Odor Odorless
Weight ≈0.18g/25x25mm
Form Foil
Assay Percent Notes (metals basis excluding Pt)
MDL Number MFCD02091738
Percent Purity 99.9%

RUO – Research Use Only